Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772573.1 | XP_008770795.1 | 20 | 2110 | ||
XM_008772574.1 | XP_008770796.1 | 20 | 2110 | |||
XM_008772575.1 | XP_008770797.1 | 20 | 2110 | |||
XM_008772576.1 | XP_008770798.1 | 20 | 2090 | |||
XM_008772577.1 | XP_008770799.1 | 20 | 2090 | |||
XM_008772578.1 | XP_008770800.1 | 19 | 2052 | |||
XM_008772579.1 | XP_008770801.1 | 19 | 2032 | |||
XM_008772580.1 | XP_008770802.1 | 19 | 2032 | |||
XM_008772581.1 | XP_008770803.1 | 20 | 2110 | |||
XM_008772582.1 | XP_008770804.1 | 19 | 2032 |