Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100850.1 | NP_001094320.1 | 3 | 1620 | ||
XM_006233918.2 | XP_006233980.1 | 3 | 417 | |||
GenBank mRNA | DQ294401.1 | - | 3 | 362 | ||
DQ294402.1 | - | 3 | 362 |
Gene Symbol: LOC100909750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749439.3 | XP_003749487.1 | 417 |