Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC686967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749071.3 | XP_003749119.2 | 4 | 941 | ||
XM_008760279.1 | XP_008758501.1 | 3 | 1088 |