Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303264.1 | NP_001290193.1 | 3 | 3165 | ||
NM_014779.3 | NP_055594.1 | 4 | 3237 | |||
NR_130136.1 | 5 | 3394 | ||||
GenBank mRNA | AB014569.1 | - | 4 | 3237 | ||
AF201290.1 | - | 4 | 593 | |||
AF201291.1 | - | 3 | 292 | |||
AF201292.1 | - | 5 | 550 | |||
AK297383.1 | - | 4 | 1059 | |||
AK297748.1 | - | 3 | 494 | |||
BC044643.1 | - | 4 | 2571 | 5454441 |