Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303264.1 | NP_001290193.1 | 1 | 2949 | ||
NM_014779.3 | NP_055594.1 | 1 | 2949 | |||
NR_130136.1 | 1 | 2949 | ||||
XM_011513337.2 | XP_011511639.1 | 1 | 3016 | |||
GenBank mRNA | AB014569.1 | - | 1 | 2949 | ||
AF201290.1 | - | 1 | 305 | |||
AF201291.1 | - | 1 | 76 | |||
AF201292.1 | - | 1 | 105 | |||
AK297383.1 | - | 2 | 843 | |||
AK297748.1 | - | 1 | 278 | |||
BC044643.1 | - | 1 | 2283 | 5454441 |