Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ajap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101014.1 | NP_001094484.1 | 2 | 1061 | ||
GenBank mRNA | AB179740.1 | - | 2 | 1061 |