Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100913063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228537.2 | XP_006228599.2 | 2 | 294 | ||
XM_008759062.1 | XP_008757284.1 | 2 | 595 |
Gene Symbol: Cd79a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001060872.5 | XP_001060872.3 | 2 | 285 |