Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014788.3 | NP_055603.2 | 3 | 529 | ||
NM_033219.2 | NP_150088.1 | 3 | 529 | |||
XM_005252320.3 | XP_005252377.1 | 3 | 594 | |||
XM_006717338.1 | XP_006717401.1 | 3 | 1152 | |||
XM_011519244.2 | XP_011517546.1 | 3 | 359 | |||
XM_017015352.1 | XP_016870841.1 | 3 | 594 | |||
XM_017015353.1 | XP_016870842.1 | 3 | 594 | |||
XM_017015354.1 | XP_016870843.1 | 3 | 585 | |||
GenBank mRNA | AF220130.1 | - | 3 | 374 | ||
AF220131.1 | - | 3 | 374 | |||
AK055833.1 | - | 3 | 402 | |||
AK292825.1 | - | 3 | 396 | |||
BC006333.2 | - | 3 | 382 | 4299815 | ||
BF797334.1 | - | 3 | 487 | |||
D50919.1 | - | 3 | 374 | |||
DC405577.1 | - | 3 | 529 |