Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1561671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_341496.7 | XP_341497.4 | 1 | 253 |
Gene Symbol: Uqcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108528.1 | NP_001101998.1 | 3 | 294 | ||
XM_006256124.2 | XP_006256186.1 | 4 | 319 | |||
XM_008772733.1 | XP_008770955.1 | 4 | 743 | |||
GenBank mRNA | BC169127.1 | - | 461 | 6918056 |