Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082480.2 | NP_001075949.1 | 2 | 893 | ||
NM_001261843.1 | NP_001248772.1 | 2 | 772 | |||
NM_014789.3 | NP_055604.3 | 1 | 753 | |||
XM_006716708.3 | XP_006716771.1 | 2 | 825 | |||
GenBank mRNA | AB014528.1 | - | 1 | 877 | ||
AK294695.1 | - | 2 | 765 | |||
BC067516.1 | - | 1 | 753 | 7002215 | ||
BC067517.1 | - | 1 | 753 | 7002216 | ||
BC067518.1 | - | 1 | 753 | 7002218 | ||
BC075056.2 | - | 1 | 702 | 30915366 | ||
BC075057.2 | - | 1 | 700 | 30915582 | ||
BC112899.1 | - | 1 | 267 |