Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169139.1 | NP_001162610.1 | 7 | 1500 | ||
GenBank mRNA | EV778235.1 | - | 4 | 459 |