Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270934.1 | NP_001257863.1 | 3 | 808 | ||
NM_001270941.1 | NP_001257870.1 | 3 | 808 | |||
NM_001282282.1 | NP_001269211.1 | 2 | 531 | |||
NM_014790.4 | NP_055605.2 | 3 | 808 | |||
XM_017010097.1 | XP_016865586.1 | 4 | 802 | |||
XM_017010098.1 | XP_016865587.1 | 3 | 808 | |||
GenBank mRNA | AB011127.1 | - | 3 | 502 | ||
AF273057.1 | - | 3 | 649 | |||
AK291280.1 | - | 3 | 530 | |||
AK299724.1 | - | 2 | 457 | |||
BC003189.1 | - | 3 | 454 | |||
BC017354.2 | - | 3 | 573 | 5020741 | ||
EF512550.1 | - | 2 | 267 |