Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021067.4 | NP_066545.3 | 3 | 309 | ||
NR_134574.1 | 3 | 309 | ||||
XM_017028162.1 | XP_016883651.1 | 3 | 266 | |||
GenBank mRNA | AK294593.1 | - | 3 | 263 | ||
BC012542.1 | - | 3 | 310 | 4333095 | ||
BP290581.1 | - | 309 | ||||
D80008.1 | - | 3 | 265 |