Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eid3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044304.1 | NP_001037769.2 | 1 | 651 | ||
GenBank mRNA | BC097404.1 | - | 1 | 651 | 7453955 |