Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014797.2 | NP_055612.2 | 7 | 1854 | ||
GenBank mRNA | AB007901.1 | - | 7 | 1854 | ||
BC117374.1 | - | 6 | 1722 | 40125925 | ||
BC117376.1 | - | 6 | 1722 | 40125927 | ||
BX648883.1 | - | 1030 |