Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014798.2 | NP_055613.1 | 9 | 2923 | ||
NR_027774.1 | 8 | 2675 | ||||
NR_027782.1 | 8 | 2538 | ||||
XM_006722201.3 | XP_006722264.1 | 7 | 2704 | |||
XM_011525523.2 | XP_011523825.1 | 9 | 2894 | |||
XM_011525524.1 | XP_011523826.1 | 9 | 3073 | |||
XM_011525525.1 | XP_011523827.1 | 8 | 2681 | |||
XM_011525526.1 | XP_011523828.1 | 10 | 2717 | |||
XM_011525528.2 | XP_011523830.1 | 4 | 1267 | |||
XM_017025451.1 | XP_016880940.1 | 9 | 2871 | |||
XM_017025452.1 | XP_016880941.1 | 8 | 2626 | |||
XM_017025453.1 | XP_016880942.1 | 10 | 3080 | |||
XM_017025454.1 | XP_016880943.1 | 8 | 2771 | |||
GenBank mRNA | AB002354.2 | - | 9 | 2876 | ||
AJ002220.1 | - | 7 | 2511 | |||
AK074103.1 | - | 5 | 1671 | |||
AK299116.1 | - | 8 | 2623 | |||
AK299467.1 | - | 8 | 2677 | |||
BC064361.1 | - | 9 | 2887 | 5535600 |