Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249328.2 | XP_006249390.1 | 8 | 2017 | ||
XM_006249329.2 | XP_006249391.1 | 8 | 1987 | |||
XM_006249330.2 | XP_006249392.1 | 7 | 1932 | |||
XM_008769264.1 | XP_008767486.1 | 7 | 1939 |