Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001246665.1 | NP_001233594.1 | 2 | 82 | ||
XM_006249328.2 | XP_006249390.1 | 3 | 244 | |||
XM_006249329.2 | XP_006249391.1 | 3 | 214 | |||
XM_006249330.2 | XP_006249392.1 | 2 | 159 | |||
XM_008769264.1 | XP_008767486.1 | 2 | 166 | |||
GenBank mRNA | AB606422.1 | - | 2 | 82 | ||
DV214760.1 | - | 2 | 267 | |||
DY311222.1 | - | 1 | 35 |