Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109517.1 | NP_001102987.1 | 4 | 1172 | ||
XM_006228793.2 | XP_006228855.1 | 5 | 1844 | |||
XM_008759188.1 | XP_008757410.1 | 5 | 2197 |