Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246610.2 | XP_006246672.1 | 22, 23 | 3130 | ||
XM_006246611.2 | XP_006246673.1 | 21, 22 | 2953 | |||
XM_006246612.2 | XP_006246674.1 | 20, 21 | 2662 |