Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012296.3 | NP_036428.1 | 6 | 1601 | ||
NM_080491.2 | NP_536739.1 | 6 | 1657 | |||
XM_006718753.2 | XP_006718816.1 | 7 | 1702 | |||
XM_011545408.2 | XP_011543710.1 | 5 | 1308 | |||
GenBank mRNA | AB011143.1 | - | 6 | 1601 | ||
AB018413.1 | - | 6 | 1555 | |||
BC131711.1 | - | 6 | 1559 | 40122341 | ||
BC152459.1 | - | 6 | 1601 | 8862595 |