Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC365839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749325.3 | XP_003749373.1 | 2 | 1130 |
Gene Symbol: RGD1566052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115047.2 | NP_001108519.1 | 9 | 1083 | ||
XM_008762071.1 | XP_008760293.1 | 9 | 1138 | |||
GenBank mRNA | BC158667.1 | - | 9 | 1083 | 8370237 |