Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 16 | 2734 | ||
XM_006721612.2 | XP_006721675.1 | 14 | 2177 | |||
XM_011524090.2 | XP_011522392.1 | 17 | 2800 | |||
XM_011524091.2 | XP_011522393.1 | 17 | 4185 | |||
XM_011524095.2 | XP_011522397.1 | 17 | 2532 | |||
XM_011524096.1 | XP_011522398.1 | 17 | 2800 | |||
XM_011524098.2 | XP_011522400.1 | 15 | 2431 | |||
XM_011524099.2 | XP_011522401.1 | 15 | 1921 | |||
XM_011524100.2 | XP_011522402.1 | 14 | 1703 | |||
XM_017025455.1 | XP_016880944.1 | 17 | 2743 | |||
XM_017025456.1 | XP_016880945.1 | 14 | 2495 | |||
XM_017025457.1 | XP_016880946.1 | 13 | 2265 | |||
GenBank mRNA | AB018296.1 | - | 16 | 2479 | ||
AK292876.1 | - | 16 | 2496 | |||
AK296971.1 | - | 15 | 2376 | |||
AK300547.1 | - | 9 | 882 | |||
AL080108.1 | - | 10 | 1167 | |||
BC017566.1 | - | 16 | 2493 | |||
BC113016.1 | - | 15 | 2449 | 40036441 | ||
BC113017.1 | - | 14 | 2342 | 40036446 |