Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 17 | 2865 | ||
XM_006721612.2 | XP_006721675.1 | 15 | 2308 | |||
XM_011524090.2 | XP_011522392.1 | 18 | 2931 | |||
XM_011524091.2 | XP_011522393.1 | 18 | 4316 | |||
XM_011524095.2 | XP_011522397.1 | 18 | 2663 | |||
XM_011524096.1 | XP_011522398.1 | 18 | 2931 | |||
XM_011524098.2 | XP_011522400.1 | 16 | 2562 | |||
XM_011524099.2 | XP_011522401.1 | 16 | 2052 | |||
XM_011524100.2 | XP_011522402.1 | 15 | 1834 | |||
XM_017025455.1 | XP_016880944.1 | 18 | 2874 | |||
XM_017025456.1 | XP_016880945.1 | 15 | 2626 | |||
XM_017025457.1 | XP_016880946.1 | 14 | 2396 | |||
GenBank mRNA | AB018296.1 | - | 17 | 2610 | ||
AK292876.1 | - | 17 | 2627 | |||
AK296971.1 | - | 16 | 2507 | |||
AK300547.1 | - | 10 | 1013 | |||
AL080108.1 | - | 11 | 1298 | |||
BC113016.1 | - | 16 | 2580 | 40036441 | ||
BC113017.1 | - | 15 | 2473 | 40036446 |