Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135999.1 | NP_001129471.1 | 2 | 813 | ||
NM_014806.3 | NP_055621.1 | 2 | 557 | |||
NR_052015.2 | 2 | 557 | ||||
GenBank mRNA | AB002373.2 | - | 2 | 399 | ||
AK299805.1 | - | 2 | 503 | |||
BC132766.1 | - | 1 | 342 | 40146788 | ||
BC132770.1 | - | 1 | 342 | 40146792 | ||
BC146654.1 | - | 2 | 531 | 40147826 | ||
BC150211.1 | - | 2 | 469 | |||
BC150262.1 | - | 2 | 410 | 9007205 | ||
BX647690.1 | - | 2 | 2652 | |||
DB458531.2 | - | 2 | 501 | |||
DB460837.2 | - | 2 | 558 |