Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014808.3 | NP_055623.1 | 25 | 2970 | ||
XM_011512233.1 | XP_011510535.1 | 26 | 2955 | |||
XM_011512234.1 | XP_011510536.1 | 26 | 2880 | |||
XM_011512235.1 | XP_011510537.1 | 27 | 3213 | |||
XM_011512236.1 | XP_011510538.1 | 26 | 3009 | |||
XM_017005416.1 | XP_016860905.1 | 27 | 3170 | |||
XM_017005417.1 | XP_016860906.1 | 26 | 3149 | |||
XM_017005418.1 | XP_016860907.1 | 23 | 2787 | |||
GenBank mRNA | AB018336.1 | - | 25 | 2923 | ||
AK127510.1 | - | 4 | 2201 | |||
AL122052.1 | - | 4 | 335 |