Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282983.1 | NP_001269912.1 | 10 | 1134 | ||
NM_001282984.1 | NP_001269913.1 | 10 | 1134 | |||
NM_014808.3 | NP_055623.1 | 10 | 1134 | |||
XM_005247050.3 | XP_005247107.1 | 10 | 1098 | |||
XM_011512233.1 | XP_011510535.1 | 10 | 1098 | |||
XM_011512234.1 | XP_011510536.1 | 10 | 1023 | |||
XM_011512235.1 | XP_011510537.1 | 11 | 1356 | |||
XM_011512236.1 | XP_011510538.1 | 10 | 1152 | |||
XM_017005416.1 | XP_016860905.1 | 11 | 1313 | |||
XM_017005417.1 | XP_016860906.1 | 11 | 1313 | |||
XM_017005418.1 | XP_016860907.1 | 8 | 951 | |||
XM_017005419.1 | XP_016860908.1 | 10 | 1098 | |||
XM_017005420.1 | XP_016860909.1 | 10 | 1098 | |||
GenBank mRNA | AB018336.1 | - | 10 | 1087 | ||
AK300435.1 | - | 10 | 1094 | |||
BC021301.2 | - | 10 | 1086 | 5013180 |