Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106840.1 | NP_001100310.1 | 615 | |||
GenBank mRNA | BC157819.1 | - | 585 | 5621777 | ||
FJ179399.1 | - | 5, 6 | 548 | |||
FQ230022.1 | - | 595 | ||||
FQ233696.1 | - | 597 | ||||
FQ233840.1 | - | 597 | ||||
X62660.1 | - | 576 |
Gene Symbol: LOC100909835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750542.3 | XP_003750590.1 | 1 | 634 |