Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168374.1 | NP_001161846.1 | 6 | 1631 | ||
NM_001168375.1 | NP_001161847.1 | 5 | 1515 | |||
NM_001168376.1 | NP_001161848.1 | 4 | 1355 | |||
NM_001168377.1 | NP_001161849.1 | 5 | 1540 | |||
NM_014809.3 | NP_055624.2 | 5 | 1540 | |||
XM_011515025.2 | XP_011513327.1 | 5 | 1297 | |||
XM_011515026.2 | XP_011513328.1 | 4 | 1258 | |||
XM_017011541.1 | XP_016867030.1 | 6 | 1611 | |||
XM_017011542.1 | XP_016867031.1 | 5 | 1258 | |||
XM_017011545.1 | XP_016867034.1 | 5 | 1253 | |||
XM_017011546.1 | XP_016867035.1 | 5 | 1258 | |||
XM_017011547.1 | XP_016867036.1 | 5 | 1258 | |||
XM_017011548.1 | XP_016867037.1 | 5 | 1331 | |||
XM_017011549.1 | XP_016867038.1 | 5 | 1288 | |||
XM_017011550.1 | XP_016867039.1 | 5 | 1258 | |||
XM_017011551.1 | XP_016867040.1 | 5 | 1258 | |||
XM_017011552.1 | XP_016867041.1 | 6 | 1215 | |||
XM_017011553.1 | XP_016867042.1 | 6 | 1057 | |||
GenBank mRNA | AB002317.1 | - | 5 | 1526 | ||
AK295008.1 | - | 4 | 1092 | |||
AK296310.1 | - | 5 | 1222 | |||
AK296426.1 | - | 6 | 1366 | |||
AK307503.1 | - | 5 | 1225 | |||
BC140821.1 | - | 6 | 1371 | 9021689 | ||
BC144628.1 | - | 5 | 1399 | 9053159 | ||
BC152460.1 | - | 5 | 1526 | 8862596 |