Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014811.3 | NP_055626.3 | 4 | 3040 | ||
XM_005263411.2 | XP_005263468.1 | 4 | 2845 | |||
XM_017015355.1 | XP_016870844.1 | 4 | 3322 | |||
XM_017015356.1 | XP_016870845.1 | 4 | 3059 | |||
XM_017015357.1 | XP_016870846.1 | 4 | 2913 | |||
XM_017015358.1 | XP_016870847.1 | 4 | 4281 | |||
XM_017015359.1 | XP_016870848.1 | 4 | 3855 | |||
XM_017015360.1 | XP_016870849.1 | 4 | 3865 | |||
XM_017015361.1 | XP_016870850.1 | 3 | 3663 | |||
XM_017015362.1 | XP_016870851.1 | 3 | 3880 | |||
GenBank mRNA | AK022736.1 | - | 1 | 775 |