Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamstr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001076631.5 | XP_001076631.2 | 3 | 1044 | ||
XM_006229210.2 | XP_006229272.1 | 3 | 1429 | |||
XM_006229211.2 | XP_006229273.1 | 3 | 988 |