Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271779.1 | NP_001258708.1 | 7 | 1225 | ||
NM_001271780.1 | NP_001258709.1 | 6 | 940 | |||
NM_001271781.1 | NP_001258710.1 | 6 | 815 | |||
NM_014814.2 | NP_055629.1 | 6 | 1066 | |||
XM_005265619.1 | XP_005265676.1 | 6 | 1162 | |||
XM_017007569.1 | XP_016863058.1 | 7 | 968 | |||
GenBank mRNA | AF215935.1 | - | 6 | 932 | ||
AF530062.1 | - | 6 | 879 | |||
AK055210.1 | - | 2 | 1568 | |||
AK095059.1 | - | 5 | 1648 | |||
AK290205.1 | - | 6 | 920 | |||
AL529900.3 | - | 6 | 801 | |||
AL832109.1 | - | 1 | 4176 | |||
AY359879.1 | - | 6 | 912 | |||
AY568086.1 | - | 6 | 904 | |||
BC000630.1 | - | 6 | 896 | 3344973 | ||
BC000904.2 | - | 6 | 917 | 2900971 | ||
BC012369.1 | - | 6 | 914 | 3858094 | ||
BX647742.1 | - | 7 | 1092 | |||
D14663.1 | - | 6 | 904 |