Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271779.1 | NP_001258708.1 | 9 | 1471 | ||
NM_001271780.1 | NP_001258709.1 | 8 | 1186 | |||
NM_001271781.1 | NP_001258710.1 | 8 | 1061 | |||
NM_014814.2 | NP_055629.1 | 8 | 1312 | |||
XM_005265619.1 | XP_005265676.1 | 8 | 1408 | |||
XM_011534288.1 | XP_011532590.1 | 8 | 1198 | |||
XM_017007569.1 | XP_016863058.1 | 9 | 1214 | |||
GenBank mRNA | AF215935.1 | - | 8 | 1178 | ||
AF530062.1 | - | 8 | 1125 | |||
AK054730.1 | - | 2 | 2463 | |||
AK055210.1 | - | 4 | 1814 | |||
AK095059.1 | - | 7 | 1894 | |||
AK290205.1 | - | 8 | 1166 | |||
AL832109.1 | - | 3 | 4422 | |||
AY359879.1 | - | 8 | 1158 | |||
AY568086.1 | - | 8 | 1150 | |||
BC000630.1 | - | 8 | 1142 | 3344973 | ||
BC000904.2 | - | 8 | 1163 | 2900971 | ||
BC012369.1 | - | 8 | 1160 | 3858094 | ||
BX647742.1 | - | 9 | 1338 | |||
CD110455.1 | - | 2 | 104 | |||
CD678025.1 | - | 1 | 377 | |||
D14663.1 | - | 8 | 1150 |