Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055430.5 | XP_001055430.4 | 4, 5 | 917 | ||
XM_006241414.2 | XP_006241476.1 | 5, 6 | 511 | |||
XM_008765427.1 | XP_008763649.1 | 4, 5 | 937 | |||
XM_008765428.1 | XP_008763650.1 | 5, 6 | 561 |