Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gngt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135767.1 | NP_001129239.1 | 3, 4 | 276 | ||
XM_006247208.2 | XP_006247270.1 | 3, 4 | 387 | |||
XM_006247209.1 | XP_006247271.1 | 3, 4 | 185 | |||
XM_006247210.1 | XP_006247272.1 | 3, 4 | 185 | |||
XM_006247211.2 | XP_006247273.1 | 2, 3 | 435 | |||
XM_008768115.1 | XP_008766337.1 | 2, 3 | 332 |