Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173451.1 | NP_001166922.1 | 6 | 516 | ||
GenBank mRNA | FQ213477.1 | - | 6 | 496 | ||
FQ229682.1 | - | 6 | 496 | |||
FQ233075.1 | - | 6 | 497 |