Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039479.1 | NP_001034568.1 | 13 | 2035 | ||
XM_005268244.3 | XP_005268301.1 | 12 | 1822 | |||
XM_006720344.2 | XP_006720407.1 | 13 | 2016 | |||
XM_011537415.2 | XP_011535717.1 | 14 | 2108 | |||
XM_011537416.2 | XP_011535718.1 | 13 | 2046 | |||
XM_011537417.2 | XP_011535719.1 | 13 | 1890 | |||
GenBank mRNA | AB002315.1 | - | 13 | 2001 | ||
AK296912.1 | - | 12 | 1793 | |||
AK304217.1 | - | 13 | 2012 | |||
BC032944.2 | - | 13 | 2039 | 5267463 |