Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCHSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014824.2 | NP_055639.2 | 15, 16 | 1749 | ||
XM_011545409.1 | XP_011543711.1 | 15, 16 | 1631 | |||
XM_011545410.2 | XP_011543712.1 | 15, 16 | 1564 | |||
XM_017018632.1 | XP_016874121.1 | 15, 16 | 1808 | |||
GenBank mRNA | AB018312.1 | - | 14, 15 | 1599 | ||
AK296376.1 | - | 15, 16 | 1491 | |||
AK297928.1 | - | 15, 16 | 1633 | |||
AL133567.1 | - | 10, 11 | 1117 | |||
BC010394.1 | - | 15, 16 | 1496 | 4091973 | ||
BC017751.1 | - | 3, 4 | 299 |