Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136554.1 | NP_001130026.1 | 5, 6 | 479 | ||
NM_001136555.1 | NP_001130027.1 | 3, 4 | 250 | |||
NM_012290.4 | NP_036422.3 | 5, 6 | 920 | |||
XM_011512238.2 | XP_011510540.1 | 5, 6 | 960 | |||
XM_017005421.1 | XP_016860910.1 | 6, 7 | 1029 | |||
XM_017005422.1 | XP_016860911.1 | 4, 5 | 841 | |||
GenBank mRNA | AB004885.1 | - | 6, 7 | 723 | ||
AF162666.1 | - | 5, 6 | 601 | |||
AK090779.1 | - | 5, 6 | 479 | |||
AK091975.1 | - | 5, 6 | 920 | |||
AK292508.1 | - | 5, 6 | 561 | |||
AK301857.1 | - | 3, 4 | 250 | |||
BC032657.1 | - | 5, 6 | 481 | 5532877 | ||
D50927.2 | - | 5, 6 | 553 | |||
DA144429.1 | - | 5, 6 | 479 |