Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133297.2 | NP_579831.2 | 5 | 677 | ||
GenBank mRNA | AF390544.1 | - | 5 | 401 | ||
BC060547.1 | - | 5 | 426 | 6921948 | ||
CK596377.1 | - | 5 | 692 | |||
DY561321.1 | - | 5 | 415 | |||
FQ209574.1 | - | 5 | 412 | |||
FQ212727.1 | - | 5 | 445 | |||
FQ213177.1 | - | 5 | 426 | |||
FQ214634.1 | - | 5 | 422 | |||
FQ215992.1 | - | 5 | 435 | |||
FQ216831.1 | - | 5 | 412 | |||
FQ219237.1 | - | 5 | 433 | |||
FQ219570.1 | - | 5 | 412 | |||
FQ219725.1 | - | 5 | 405 | |||
FQ220311.1 | - | 5 | 411 | |||
FQ220593.1 | - | 5 | 417 | |||
FQ220877.1 | - | 5 | 425 | |||
FQ229133.1 | - | 5 | 418 | |||
FQ230131.1 | - | 5 | 412 |