Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022281.2 | NP_071617.2 | 7 | 772 | ||
GenBank mRNA | AF487549.1 | - | 7 | 772 | ||
AY170916.1 | - | 7 | 733 | |||
AY174892.1 | - | 7 | 772 | |||
AY174893.1 | - | 7 | 772 |
Gene Symbol: LOC100912948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245698.2 | XP_006245760.1 | 2 | 486 |