Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014831.2 | NP_055646.2 | 12 | 2949 | ||
XM_006713432.3 | XP_006713495.1 | 13 | 3117 | |||
XM_011534289.2 | XP_011532591.1 | 13 | 3878 | |||
XM_017007570.1 | XP_016863059.1 | 12 | 2749 | |||
XM_017007571.1 | XP_016863060.1 | 13 | 3010 | |||
XM_017007572.1 | XP_016863061.1 | 13 | 2912 | |||
XM_017007573.1 | XP_016863062.1 | 13 | 2920 | |||
GenBank mRNA | AB002340.3 | - | 2 | 1327 | ||
AK096678.1 | - | 1 | 565 |