Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286658.2 | NP_001273587.1 | 13 | 2933 | ||
NM_001286659.2 | NP_001273588.1 | 12 | 2768 | |||
NM_014832.4 | NP_055647.2 | 14 | 2957 | |||
XM_005266603.1 | XP_005266660.1 | 13 | 3360 | |||
XM_005266605.2 | XP_005266662.1 | 14 | 2285 | |||
XM_006719903.3 | XP_006719966.1 | 14 | 2645 | |||
XM_011535331.1 | XP_011533633.1 | 13 | 3324 | |||
XM_017020882.1 | XP_016876371.1 | 12 | 1977 | |||
XM_017020883.1 | XP_016876372.1 | 11 | 1971 | |||
XM_017020884.1 | XP_016876373.1 | 2 | 1543 | |||
GenBank mRNA | AB011175.1 | - | 14 | 2903 | ||
AB449885.1 | - | 14 | 2556 | |||
AK300468.1 | - | 3 | 466 | |||
AK304091.1 | - | 3 | 446 | |||
AY211910.1 | - | 3 | 528 | |||
BC151239.1 | - | 14 | 2895 | 8860512 | ||
CR749622.1 | - | 4 | 737 | |||
FM207106.1 | - | 12 | 2367 | |||
FM207107.1 | - | 13 | 2532 |