Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109503.1 | NP_001102973.1 | 4 | 351 | ||
GenBank mRNA | BC098743.1 | - | 3 | 920 | ||
FQ229674.1 | - | 4 | 295 |