Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109908.1 | NP_001103378.1 | 5, 6 | 474 | ||
NM_012554.3 | NP_036686.2 | 5, 6 | 425 | |||
XM_006239444.1 | XP_006239506.1 | 5, 6 | 441 | |||
GenBank mRNA | AF241613.1 | - | 28 | |||
BC063174.1 | - | 5, 6 | 418 | |||
BC078896.1 | - | 5, 6 | 474 | 7112523 | ||
BC081847.1 | - | 5, 6 | 398 | 7189453 | ||
BC090069.1 | - | 5, 6 | 355 | 7108936 | ||
BC091572.1 | - | 5, 6 | 395 | 7107492 | ||
BC097343.1 | - | 5, 6 | 391 | 7461555 | ||
FQ214707.1 | - | 5, 6 | 347 | |||
FQ225935.1 | - | 5, 6 | 438 | |||
X02610.1 | - | 5, 6 | 414 |