Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278649.1 | NP_001265578.1 | 6, 7 | 686 | ||
NM_014835.3 | NP_055650.1 | 6, 7 | 705 | |||
NM_144498.2 | NP_653081.1 | 6, 7 | 741 | |||
XM_017028163.1 | XP_016883652.1 | 6, 7 | 741 | |||
XM_017028164.1 | XP_016883653.1 | 7, 8 | 1046 | |||
XM_017028165.1 | XP_016883654.1 | 7, 8 | 1046 | |||
XM_017028166.1 | XP_016883655.1 | 7, 8 | 795 | |||
XM_017028167.1 | XP_016883656.1 | 7, 8 | 809 | |||
XM_017028168.1 | XP_016883657.1 | 6, 7 | 741 | |||
XM_017028169.1 | XP_016883658.1 | 6, 7 | 665 | |||
XM_017028170.1 | XP_016883659.1 | 6, 7 | 665 | |||
GenBank mRNA | AB018315.1 | - | 6, 7 | 611 | ||
AF392447.1 | - | 6, 7 | 702 | |||
AK000230.1 | - | 6, 7 | 702 | |||
AK093592.1 | - | 2 | 844 | |||
AK124174.1 | - | 1 | 52 | |||
AK291851.1 | - | 6, 7 | 669 | |||
AK296595.1 | - | 6, 7 | 647 | |||
AK315576.1 | - | 6, 7 | 702 | |||
AY028168.1 | - | 6, 7 | 611 | |||
BC000296.2 | - | 6, 7 | 645 | 2819652 | ||
BC004455.1 | - | 6, 7 | 645 | 2819652 | ||
BC018812.2 | - | 6, 7 | 645 | |||
CR456722.1 | - | 5, 6 | 500 |