Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047116.1 | NP_001040581.1 | 2, 3 | 473 | ||
XM_006236003.2 | XP_006236065.1 | 2, 3 | 1517 | |||
GenBank mRNA | BC107563.1 | - | 2, 3 | 473 | 7938579 |