Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kiaa0895l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044292.1 | NP_001037757.1 | 2 | 991 | ||
XM_008772533.1 | XP_008770755.1 | 2 | 1649 | |||
GenBank mRNA | BC098045.1 | - | 1 | 562 | 7457085 | |
BC098942.1 | - | 2 | 991 | 7456844 |