Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114735.1 | NP_001108207.1 | 1 | 531 | ||
NM_004049.3 | NP_004040.1 | 1 | 531 | |||
GenBank mRNA | AY234180.1 | - | 1 | 357 | ||
BC016281.1 | - | 1 | 409 | 3920808 | ||
BG198875.1 | - | 1 | 677 | |||
BG216703.1 | - | 1 | 532 | |||
BT007103.1 | - | 1 | 349 | |||
CD640106.1 | - | 1 | 297 | |||
CR541937.1 | - | 1 | 349 | |||
CR541962.1 | - | 1 | 349 | |||
U27467.1 | - | 1 | 383 | |||
U29680.1 | - | 1 | 427 | |||
Y09397.1 | - | 1 | 451 |