Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033117.2 | NP_001028289.1 | 6 | 1492 | ||
NM_014850.3 | NP_055665.1 | 6 | 1492 | |||
XM_011534295.1 | XP_011532597.1 | 6 | 1492 | |||
XM_011534296.1 | XP_011532598.1 | 7 | 1609 | |||
XM_011534301.2 | XP_011532603.1 | 3 | 3321 | |||
XM_011534303.1 | XP_011532605.1 | 3 | 335 | |||
XM_017007574.1 | XP_016863063.1 | 6 | 1830 | |||
XM_017007575.1 | XP_016863064.1 | 6 | 1830 | |||
XM_017007576.1 | XP_016863065.1 | 8 | 999 | |||
XM_017007577.1 | XP_016863066.1 | 8 | 938 | |||
XM_017007578.1 | XP_016863067.1 | 6 | 1858 | |||
XM_017007579.1 | XP_016863068.1 | 6 | 1826 | |||
XM_017007580.1 | XP_016863069.1 | 8 | 1059 | |||
GenBank mRNA | AB007871.2 | - | 6 | 716 | ||
AB032982.1 | - | 4 | 367 | |||
AF427144.1 | - | 6 | 860 | |||
AF464189.1 | - | 6 | 1020 | |||
AK094355.1 | - | 6 | 798 | |||
BC039300.1 | - | 6 | 827 |